Spectrophometry Cuvettes
Spectrophometry Cuvettes
Actual product may vary
Fisherbrand
Spectrophometry Cuvettes
Manufactured with optical polystyrene or UV grade PMMA for clear and sharp optical path with indication of optical path direction. Kartell™ Spectrophometry Cuvettes are ideal for spectrophotometry application.
Have Questions?
Catalog NumberCapacity (Metric)Wavelength RangeType
FKS107817912.5 mL340 to 800 nmSemi-micro Cell
Catalog number FKS10781791
Price (MYR)
134.00
PK
Add to cart
Capacity (Metric):
2.5 mL
Wavelength Range:
340 to 800 nm
Type:
Semi-micro Cell
Price (MYR)
134.00
PK
Add to cart
Manufactured with optical polystyrene or UV grade PMMA for clear and sharp optical path with indication of optical path direction. Kartell™ Spectrophometry Cuvettes are ideal for spectrophotometry application.
  • Equipped with optically machined walls (optical faces) to ensure optimal transparency along the spectral field
    • Polystrene range:340nm to 800nm
    • UV range: 280 to 800nm
  • Path length 10mm
  • Compatible with most spectrophotometers and photometers on the market
  • Absorption changes: within the ±1% interval
  • CE marked--IVD approved
Specifications
TypeSemi-micro Cell
DisposableSingle-use
Capacity (Metric)2.5 mL
MaterialPolystyrene
Length (Metric)12mm
Certifications/ComplianceCE-IVD
Quantity100Pack
Path Length10 mm
Wavelength Range340 to 800 nm
Height (Metric)45 mm
For Use With (Equipment)Spectrophotometry
Width (Metric)12 mm
For Use With (Application)Spectrophotometry
Unit SizePK