Round-Bottom Polystyrene Test Tubes
Round-Bottom Polystyrene Test Tubes
Actual product may vary
Round-Bottom Polystyrene Test Tubes
Constructed of optically clear polystyrene for easy viewing of sample. Falcon™ Round-Bottom Polystyrene Tubes are widely used and referenced in laboratory protocols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure TypeSterilityPackaging
FKS1076986116 mLScrew CapSterileBag of 125, 1000/Cs.
CRG35204519 mLScrew CapSterileIndividually wrapped, 500/Cs.
CRG3520545 mLSnap CapSterileBag of 125, 1000/Cs.
FIS14-959-2A5 mLSnap CapSterileBag of 125, 1000/Cs.
CRG3520525 mLNot IncludedSterile1000/Cs.
CRG3520035 mLSnap CapSterileIndividually wrapped, 500/Cs.
Catalog number FKS10769861
Price (MYR)
4,903.00
PK
Add to cart
Capacity (Metric):
16 mL
Closure Type:
Screw Cap
Sterility:
Sterile
Packaging:
Bag of 125, 1000/Cs.
Price (MYR)
4,903.00
PK
Add to cart
  • Widely used and referenced in laboratory protocols
  • Dual position snap-cap prevents sample loss with ease of handling; provides a secure, positive seal (except for items 352008, 352017, and 352052 which do not have caps) 
  • Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352008 nonsterile)
The foundation for consistent research results
  • Provide reliable containment of laboratory fluid samples
  • Polystyrene tubes are best suited for procedures requiring high optical clarity
  • Heavier gauge walls; unique construction 
  • Medical-style packaging materials
  • 1,400 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
  • Tube dimensions and volumes are approximate
Specifications
Closure ColorBlack
ColorClear
Description1400 x G
Capacity (Metric)16 mL
Length (Metric)125 mm
MaterialPolystyrene
DisposableSingle-use
IncludesDual position snap cap
PackagingBag of 125, 1000/Cs.
SterilitySterile
GraduatedNo
Diameter (Metric)16 mm
Closure TypeScrew Cap
Product TypeTest Tube
Unit SizePK