Check Order Status
Soda Lime Glass Short Stem Funnel
Actual product may vary
Fisherbrand
Soda Lime Glass Short Stem Funnel
Manufactured from soda lime glass. Fisherbrand™ Soda Lime Glass Short Stem Funnel is less likely to clog when funneling thicker substances.
Have Questions?
Change view
Catalog Number
Diameter (Metric)Top
Diameter (Metric) Stem
Length (Metric) Stem
FKS10767414
80mm
9mm
85mm
FKS10123682
100mm
12mm
110mm
2 Options
Catalog number
FKS10767414
Price (MYR)
512.00
PK
Add to cart
Diameter (Metric)Top:
80mm
Diameter (Metric) Stem:
9mm
Length (Metric) Stem:
85mm
Price (MYR)
512.00
PK
Add to cart
Manufactured from Soda Lime glass.
Short stem funnel
Less likely to clog when funneling thicker substances.
Specifications
Diameter (Metric)Top
80mm
Diameter (Metric) Stem
9mm
Material
Soda Lime Glass
Height (Metric)
140 mm
For Use With (Application)
Ideal for filtering applications in the laboratory
Length (Metric) Stem
85mm