Check Order Status
BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Actual product may vary
Fisherbrand
BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Calibrated density bottle made with Borosilicate glass that is ideal for use in most research, scientific, and other types of laboratory environments.
Have Questions?
Catalog Number
Capacity (Metric)
FKS10747981
25 mL
Catalog number
FKS10747981
Price (MYR)
854.00
EA
Add to cart
Capacity (Metric):
25 mL
Price (MYR)
854.00
EA
Add to cart
Calibrated density bottle made with Borosilicate glass
DIN ISO 3507, type Gay-Lussac
Calibrated to contain (TC, In)
Includes individual certificate
Stopper NS 10/19 with capillary
Top of stopper ground and polished
The volume in cm³ is specified to a precision of 3 decimal places
Specifications
Quantity
1Each
Type
Gay-Lussac
Capacity (Metric)
25 mL
Material
Borosilicate Glass
Product Type
Pycnometer