Mucasol™ Universal Detergent Liquid Concentrate
Mucasol™ Universal Detergent Liquid Concentrate
Actual product may vary
Mucasol™ Universal Detergent Liquid Concentrate
Cleans instruments made of glass, plastic, rubber, and metal in 10 to 30 minutes without mechanical help. BRAND™ Mucasol™ Universal Detergent Liquid Concentrate replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, nonaggressive and biodegradable.
Have Questions?
Catalog NumberVolume (Metric)Container Type
FKS107293012 LBottle
Catalog number FKS10729301
Price (MYR)
-
Volume (Metric):
2 L
Container Type:
Bottle
  • Liquid, alkaline high-performance concentrate for immersion and ultrasonic baths. Universal cleaner for laboratory equipment and instruments made of glass, porcelain, plastic, rubber and metal
  • Items are cleaned in about 10 to 30 minutes, without mechanical help
  • Mucasol™ replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, nonaggressive and biodegradable
  • Prolonged exposure of zinc and non-anodized aluminum surfaces should be avoided
Specifications
Product TypeMucasol Universal Detergent
pH∼11.5 (0.7%), ∼11.7 (3%)
AppearanceLight Green
CompositionAlkaline; >30% phosphates, <5% anionic surfactants, amphoteric surfactants, complexing agents,
ConcentrationUltrasonic bath: 0.2 to 2% (1.4 to 14 mL/L); Immersion bath: 0.7 to 3% (5 to 22 mL/L)
Container TypeBottle
For Use With (Application)For immersion and ultrasonic baths. Universal cleaner for laboratory equipment and instruments made of glass, porcelain, plastic, rubber and Metal
Quantity5/Pk.
Volume (Metric)2 L
Unit SizeEA