PTFE Beaker with Spout
PTFE Beaker with Spout
Actual product may vary
Fisherbrand
PTFE Beaker with Spout
Includes a machined base for good heat transfer. The PTFE beaker includes a molded-in spout for easy pouring.
Have Questions?
Catalog NumberCapacity (Metric)
FKS10713101250 mL
Catalog number FKS10713101
Price (MYR)
224.00
EA
Add to cart
Capacity (Metric):
250 mL
Price (MYR)
224.00
EA
Add to cart
Non-contaminating and inert to highly corrosive reagents.
  • Non-wettable, lightweight, and heat resistant up to 280°C
  • Machined base for good heat transfer
  • Easy to clean
  • Molded-in spout for easy pouring
  • Isostatically molded from pure PTFE
Specifications
ColorWhite
Diameter (Metric) Outer66mm
Diameter (Metric)Top66mm
Capacity (Metric)250 mL
MaterialPTFE
Height (Metric)97mm
Temperature (Metric) Max. Operating280°C
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet
Unit SizeEA