Rubber Tubing for Oil-Free Diaphragm Vacuum Pumps
Rubber Tubing for Oil-Free Diaphragm Vacuum Pumps
Actual product may vary
Fisherbrand
Rubber Tubing for Oil-Free Diaphragm Vacuum Pumps
For VACUUBRAND™ and Diaphragm pumps, and other pumps having a 10mm (3/8 in.) hose barb
Have Questions?
Catalog NumberOverall Length (Metric)
FKS106843323 m
Catalog number FKS10684332
Price (MYR)
777.00
EA
Add to cart
Overall Length (Metric):
3 m
Price (MYR)
777.00
EA
Add to cart
  • Inner diameter 10 mm
  • Three meter length
  • Thick walls resist collapsing
  • Made of high-quality caoutchouc rubber
Specifications
AutoclavableAutoclavable
Diameter (English) Inner0.4 in.
Diameter (Metric) Inner11 mm
MaterialCaoutchouc Rubber
Overall Length (English)9.8 ft.
Overall Length (Metric)3 m
For Use WithBrandTech pump models: ME1C, MEC, MZ1C, ME 2 and ME 4 NT Pumps
Packaging Quantity3-meter length
ColorNatural
FlexibilityFlexible
Product TypeTubing
Unit SizeEA