AR-Glas™ Stirring Rods
AR-Glas™ Stirring Rods
Actual product may vary
Fisherbrand
AR-Glas™ Stirring Rods
Glass stirring rods
Have Questions?
Catalog NumberLength (Metric)
FKS10672242200 mm
Catalog number FKS10672242
Price (MYR)
420.00
PK
Add to cart
Length (Metric):
200 mm
Price (MYR)
420.00
PK
Add to cart
Specifications
DescriptionFused closely at both ends
Diameter (Metric)5 mm
Length (Metric)200 mm
Unit SizePK