Nalgene™ Unwire™ Half-Racks
Nalgene™ Unwire™ Half-Racks
Actual product may vary
Nalgene™ Unwire™ Half-Racks
Space-efficient half-racks are ideal for use in incubators, refrigerators, freezers and under fume hoods.
Have Questions?
Catalog NumberColorHolds
FKS10549461WhiteTubes (16 mm)
Catalog number FKS10549461
Price (MYR)
214.00
EA
Add to cart
Color:
White
Holds:
Tubes (16 mm)
Price (MYR)
214.00
EA
Add to cart
Fit easily in ice or water baths without floating.
  • Made of durable ResMere™ performance plastic
  • Withstand most lab chemicals and temperatures up to 121°C (249.8°F)
  • Repeatedly autoclavable
  • Each rack holds 36 tubes
Specifications
Product TypeTest Tube Half Rack
AutoclavableAutoclavable
ColorWhite
HoldsTubes (16 mm)
MaterialPlastic
Positions36
Dimensions (L x W x H)5 x 5 x 2.7 in. (12.7 x 12.7 x 6.8 cm)
Length (Metric)12.7 cm
Length (English)5 in.
Width (Metric)12.7 cm
Width (English)5 in.
Height (Metric)6.8 cm
Height (English)2.7 in.
Diameter (Metric) Opening16 mm
Unit SizeEA