Polypropylene Round Bottom Flask Holders
Polypropylene Round Bottom Flask Holders
Actual product may vary
Fisherbrand
Polypropylene Round Bottom Flask Holders
Manufactured with polypropylene for strength and heat resistance. Kartell™ Polypropylene Round Bottom Flask Holders safely support hot flasks without damaging the holder.
Have Questions?
Catalog NumberDiameter (Metric) Outer
FKS10479022160 mm
Catalog number FKS10479022
Price (MYR)
124.00
EA
Add to cart
Diameter (Metric) Outer:
160 mm
Price (MYR)
124.00
EA
Add to cart
Manufactured with polypropylene for strength and heat resistance. Kartell™ Polypropylene Round Bottom Flask Holders safely support hot flasks without damaging the holder.
  • Internal steps provide support for a different sized flasks up to 10L
  • Withstands hot flasks up to 100°C (120°C for short periods)
  • Autoclavable
Specifications
Product TypeRound Bottom Flask Holder
AutoclavableAutoclavable
MaterialPolypropylene
ShapeRound
No. per Pack1 Pack
Quantity1 Pack
Height (Metric)49 mm
Diameter (Metric) Outer160 mm
Unit SizeEA