Nalgene™ Unitary™ LDPE Wash Bottles
Nalgene™ Unitary™ LDPE Wash Bottles
Actual product may vary
Fisher Catalogue
Nalgene™ Unitary™ LDPE Wash Bottles
Dispense contents without tipping or shaking by using these LDPE wash bottles, designed with a wide mouth for faster, easier filling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS10464301250 mL24 mm
2402-0250
also known as NLG2402-0250
250 mL24 mm
2402-0500500 mL38 mm
FIS03-409-10E500 mL38 mm
Catalog number FKS10464301
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Closure Size:
24 mm
Dispense contents without tipping or shaking with Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles. Wide mouth allows faster, easier filling. Bottle and tubulation are molded in one piece for uniform service and precision flow without sputtering. The tip can be cut back to increase flow. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. Naturally flexible LDPE material is chemically compatible with a wide range of laboratory reagents and provides an ergonomic soft squeeze to help reduce hand fatigue.
Features:
  • Bottle and tubulation are molded in one piece for uniform service
  • Tip delivers a uniform, ultrafine stream without sputtering, but can also be cut back to increase flow
  • Bottle and Carton are recyclable in many communities
  • Manufactured from non-toxic materials
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Polypropylene screw closure
Includes:
Polypropylene screw closure.
Recycling information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
ColorTranslucent
DescriptionLDPE bottle/tubulation
MaterialLDPE
No. per Case36
No. per Pack4
Closure IncludedYes
GraduatedNo
MouthWide
ShapeCylindrical
Height (Metric)137.7 mm
Product TypeWash Bottle
Height (English)5.42 in.
Closure MaterialPolypropylene
Closure Size24 mm
Diameter (Metric)62.5 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizePK