Porcelain Buchner Funnel
Porcelain Buchner Funnel
Actual product may vary
Porcelain Buchner Funnel
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) Plate
FKS1043328150 mL48mm
Catalog number FKS10433281
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
50 mL
Diameter (Metric) Plate:
48mm
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
  • Vertical sides
  • Available in a variety of sizes to fit many filtration needs
  • Perforated fixed filter plate
Specifications
Capacity (Metric)50 mL
MaterialPorcelain
Diameter (Metric) Plate48mm
Quantity1Pack
Product TypeBuchner Funnel
Unit SizeEA