Filter Membranes
Filter Membranes
Actual product may vary
Filter Membranes
Used with seripettor™ / QuikSip™ / accu-jet™ pro macro pipet controllers. Filter Membranes are available in 3μL capacity and provided in blister pack packaging.
Have Questions?
Catalog NumberPore Size
FKS103735813 μm
Catalog number FKS10373581
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Pore Size:
3 μm
  • For seripettor™ / QuikSip™ / accu-jet™ pro / macro pipette controller
  • Blister pack packaging
  • 3μm
  • Non-sterile
Specifications
MaterialPolypropylene, PTFE
Pore Size3 μm
For Use With (Application)Reduces cross-contamination, protects against over aspiration and allows pipetting to Class A tolerances
For Use With (Equipment)Macro and Accu-jet™ pipet controllers
Unit SizeEA