DURAN™ Filter Apparatus, Witt Type, complete, with interchangeable lid, and KECK™ assembly set, standard ground joint 29/32
DURAN™ Filter Apparatus, Witt Type, complete, with interchangeable lid, and KECK™ assembly set, standard ground joint 29/32
Actual product may vary
DURAN™ Filter Apparatus, Witt Type, complete, with interchangeable lid, and KECK™ assembly set, standard ground joint 29/32
DURAN™ Filter Apparatus, Witt Type, complete, with interchangeable lid, and KECK™ assembly set, standard ground joint 29/32
DURAN™ Filter Apparatus, Witt Type, complete, with interchangeable lid, and KECK™ assembly set, standard ground joint 29/32
The Witt filtration apparatus is used for solid-liquid filtration in which the filtrate is subsequently further processed.
Have Questions?
Catalog NumberDiameter (Metric)
FKS10323591100 mm
Catalog number FKS10323591
Price (MYR)
3,984.00
EA
Add to cart
Diameter (Metric):
100 mm
Price (MYR)
3,984.00
EA
Add to cart
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
  • Tight closure due to standard ground joint
  • With interchangeable lid and KECK™ assembly set
  • Fired-on, highly durable white print
Specifications
AutoclavableAutoclavable
MaterialBorosilicate Glass (3.3), PBT, Polypropylene, VMQ, EPDM
For Use With (Application)Filtering, Preferably collecting the filtrate, Filtration at reduced pressure, Vacuum filtration, Safe working under vacuum
Quantity1 Each
Standard Taper JointNS 29/32
Diameter (Metric)100 mm
Unit SizeEA