Polystyrene Disposable Culture Tubes
Polystyrene Disposable Culture Tubes
Actual product may vary
Fisher Catalogue
Polystyrene Disposable Culture Tubes
Manufactured from optically clear polystyrene for optimum clarity. Corning™ Polystyrene Disposable Culture Tubes are provided with threaded plug seal caps to prevent leakage.
Have Questions?
Catalog NumberPackaging QuantityProduct TypeIncludes
FKS1027629150/Pk., 10 Pk./Cs.Treated Culture TubePlug-seal screw cap, rack
Catalog number FKS10276291
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Packaging Quantity:
50/Pk., 10 Pk./Cs.
Product Type:
Treated Culture Tube
Includes:
Plug-seal screw cap, rack
  • Manufactured from optically clear polystyrene
  • Threaded plug seal caps prevent leakage
  • Tissue culture-treated tubes supplied racked
  • Untreated tubes supplied bulk packed
  • Sterile
  • Nonpyrogenic
Specifications
Closure MaterialLDPE
Diameter (Metric)16 mm
Product TypeTreated Culture Tube
Closure ColorOrange
Closure TypePlug Seal
Length (Metric)125 mm
MaterialPolystyrene
IncludesPlug-seal screw cap, rack
No. per Pack50/Pk.
No. per Case500/Cs.
Packaging Quantity50/Pk., 10 Pk./Cs.
SterilitySterile
Capacity (Metric)15 mL
Unit SizePK