Quickfit™ Screwthread Adapter to Fit Cone
Quickfit™ Screwthread Adapter to Fit Cone
Actual product may vary
Fisherbrand
Quickfit™ Screwthread Adapter to Fit Cone
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
Have Questions?
Catalog NumberThread SizeCone Size
FKS101879881314/23
Catalog number FKS10187988
Price (MYR)
720.00
PK
Add to cart
Thread Size:
13
Cone Size:
14/23
Price (MYR)
720.00
PK
Add to cart
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
  • With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
  • Ideal for securing thermometers, gas inlet tubes, etc
Specifications
Cone Size14/23
For Use With (Application)Ideal for securing thermometers, gas inlet tubes
IncludesPlastic cap, Silicone Rubber ring and PTFE washer
MaterialBorosilicate Glass
Product TypeCone screw thread adapter
Thread Size13
Unit SizePK