DURAN™ Filter Disc Sintered Glass, Porosity 0, edge not fused
Actual product may vary
Fisherbrand
DURAN™ Filter Disc Sintered Glass, Porosity 0, edge not fused
A prerequisite for successful work with glass filters is the selection of the right porosity. It is important to select a filter whose largest pore size is slightly below the grain size of the smallest particles still to be separated.
Have Questions?
Catalog NumberDiameter (Metric)Top
FKS1008486080 mm
Catalog number FKS10084860
Price (MYR)
399.00
Each
Diameter (Metric)Top:
80 mm
Price (MYR)
399.00
Each
Glass powder with a certain particle size distribution results in a porous, solid body. These bodies are used as filter material in the form of plates and candles. Made of DURAN™ (borosilicate glass 3.3). They are ideally suited for separations with strong acids or alkalis, for example, and thus offer advantages over other materials such as plastic or paper. Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99. Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar.
  • Edge non-melted and centered, surfaces untreated
  • Successful work through the correct choice of porosity (The largest pore size should be slightly below the grain size of the smallest particles still to be separated).
  • Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99.
  • Glass articles for further processing
Specifications
MaterialBorosilicate Glass (3.3)
For Use With (Application)Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar
No. per Pack1/Pk.
Diameter (Metric)Top80 mm
Unit SizeEach