Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Actual product may vary
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom ShapeCapacity (Metric)Thread StyleStyle
FKS10070731Round (Self-standing)3.6 mLInternalWriting Surface, A-R Base
375418Round (Self-standing)1.8 mLExternalWriting Surface, A-R Base
377267
also known as NNC377267
Round (Self-standing)1.8 mLInternalWriting Surface, A-R Base
379189Round (Self-standing)3.6 mLInternalWriting Surface, A-R Base
363452Round4.5 mLInternalWriting Surface
Catalog number FKS10070731
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Bottom Shape:
Round (Self-standing)
Capacity (Metric):
3.6 mL
Thread Style:
Internal
Style:
Writing Surface, A-R Base
Store samples from general cold storage to the vapor phase of liquid nitrogen with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, available with either internal or external threads.
Features:
  • Non-cytotoxic, non-mutagenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
Specifications
Diameter (Metric)12.5 mm
Product TypeCryogenic Storage Tube
DescriptionWith writing surface
Diameter (English) Outer0.492 in.
Capacity (Metric)3.6 mL
Bottom ShapeRound (Self-standing)
MaterialPolypropylene
Thread StyleInternal
Length (English)2.835 in.
Length (Metric)72 mm
No. per Pack400
StyleWriting Surface, A-R Base
RNase FreeYes
Vapor LN2 (to -150°C)Yes
Product LineNunc™
SterilitySterile
Anti-RotationYes
Packaging Quantity400/Pk.
Closure MaterialPolypropylene
Capacity (English)0.1217 oz.
Dnase FreeYes
Closure TypeScrew Cap
PrintedYes
Unit SizePK