UV-Cuvette Disposable Cuvets
UV-Cuvette Disposable Cuvets
Actual product may vary
Fisher Catalogue
UV-Cuvette Disposable Cuvets
Fits most spectrophotometers without adapters.
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)QuantityWidth (Metric)
FKS100467312.5 to 4.5 mL35 mm100 Pack10 mm
Catalog number FKS10046731
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2.5 to 4.5 mL
Height (Metric):
35 mm
Quantity:
100 Pack
Width (Metric):
10 mm
BrandTech™ BRAND™ UV-Cuvette Disposable Cuvets provide transparency from 230 to 900nm and eliminate cleaning and risk of cross-contamination. Ideal for DNA, RNA and protein analysis. Compatible with with most polar organic solvents, acids, and bases.
Advanced cuvet design includes recessed windows to protect against scratching in the optical pathway.
  • Disposable; cleaning and cross-contamination risk are eliminated
  • Ideal for DNA, RNA, and protein analysis
  • Chemical resistant; can be used with most polar organic solvents, acids, and bases
  • Also compatible with acetone, butanone, DMF, and concentrated hydrochloric acid
  • Grouped by manufacturing mold cavity to ensure lowest variation in extinction coefficient
  • Fits most spectrophotometers without adapters
Ultramicro size
  • Available with 8.5 and 15mm window heights (Z-dimension)
  • May be used with sample volumes as small as 70μL
  • Can be fitted with plug seal caps (sold separately) for sample storage
  • Available individually wrapped, DNA-, RNase-free
Semimicro size
  • For sample volumes between 1.5 and 3.0mL
  • Available individually wrapped, DNA-, RNase-free
Macro size
  • Holds 2.5 to 4.5mL
Specifications
TypeCuvette
DisposableSingle-use
Capacity (Metric)2.5 to 4.5 mL
MaterialPlastic
Quantity100 Pack
Path Length10 mm
Wavelength Range230 to 900 nm
Height (Metric)35 mm
For Use With (Equipment)Spectrophotometers and Photometers
Width (Metric)10 mm
For Use With (Application)DNA, RNA, Protein Analysis, Molecular Biology
Unit SizePK