TechnaCurv™ Full-Body Vest-Style Harnesses
TechnaCurv™ Full-Body Vest-Style Harnesses
Actual product may vary
TechnaCurv™ Full-Body Vest-Style Harnesses
Made of soft, flexible webbing
Have Questions?
Catalog NumberBuckle Style (Leg)D-Ring LocationIncludesSize
FKS10041591TongueBackQwik-fit chest strap, manualStandard
Catalog number FKS10041591
Price (MYR)
2,077.00
PK
Add to cart
Buckle Style (Leg):
Tongue
D-Ring Location:
Back
Includes:
Qwik-fit chest strap, manual
Size:
Standard
Price (MYR)
2,077.00
PK
Add to cart
Optional Secure-Fit leg buckles with positive engagement windows allow you to see buckle is properly engaged—no need to listen for a click on a noisy site.
  • Soft, flexible curvilinear torso webbing moves with the wearer for improved comfort and a secure fit
  • Webbing curves to fit around the neck, so it will not chafe
  • Lightweight polymer Y-back D-locator pad allows complete adjustment of back D-ring for proper fit
  • Durable visco-elastic shoulder and sub-pelvic pads permit air to circulate; keep wearer cool and comfortable
  • Single-hand operation for easy donning and doffing
  • Feature a standard Qwik-Fit™ chest strap
Specifications
Buckle StyleTongue
Buckle Style (Leg)Tongue
Capacity (English)400 lb.
Certifications/ComplianceOSHA, ANSI Z359.1 and A10.32, CSA Z259.10. Certified according to EN 361and EN 358
D-Ring LocationBack
For Use With (Application)Confined space, construction
IncludesQwik-fit chest strap, manual
SizeStandard
StyleVest
Webbing MaterialTeflon-coated Polyester
ColorRed
Product TypeFull-body harness
Unit SizePK