Polypropylene Water Jet Pump
Polypropylene Water Jet Pump
Actual product may vary
Fisherbrand
Polypropylene Water Jet Pump
Evacuates a 5L container to 20mbar in about 19min, water pressure 100kPa. Kartell™ Polypropylene Water Jet Pump works on pressures up to 1,000kPa.
Have Questions?
Catalog number FKS10020991
Price (MYR)
202.00
EA
Add to cart
Price (MYR)
202.00
EA
Add to cart
Evacuates a 5L container to 20mbar in about 19min, water pressure 100kPa. Kartell™ Polypropylene Water Jet Pump works on pressures up to 1,000kPa.
  • Non-return valve eliminates back flow
  • Useful for sucking to waste any chemicals or for filter work
  • Easily dismantled for cleaning
  • Evacuating nozzle tapers from 9 to 16mm.
  • The accessory connectors can be pushed onto the evacuating tubing to act as a quick release from the pump
  • Autoclavable
Specifications
Max. Vacuum20 mbar
MaterialPolypropylene
ApplicationsFiltration
Maximum Pressure980.66 kpa
Unit SizeEA