18/10 Stainless Steel Chattaway Double Spatulas
18/10 Stainless Steel Chattaway Double Spatulas
Actual product may vary
Fisherbrand
18/10 Stainless Steel Chattaway Double Spatulas
Have Questions?
Catalog NumberLength (Metric)Length (Metric) Blade
FKS10020801235 mm70 mm
Catalog number FKS10020801
Price (MYR)
48.00
EA
Add to cart
Length (Metric):
235 mm
Length (Metric) Blade:
70 mm
Price (MYR)
48.00
EA
Add to cart
Specifications
Quantity1 Ea.
Product TypeChattaway Double Spatula
Length (Metric) Blade70 mm
Width (Metric)10 mm
Width (Metric) Blade10 mm
Blade MaterialStainless Steel (18/10)
Diameter (Metric) Outer4mm
Dimensions (L x W)235 x 10 mm
MaterialStainless Steel (18/10)
Length (Metric)235 mm
Unit SizeEA