Check Order Status
Xylene, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Xylene, Certified AR for Analysis, Fisher Chemical™
C8H10, CAS Number-1330-20-7, 2.5L, 136 deg.C, Colorless, 106.17g/mol, -34 deg.C, 77264, Amber glass bottle, Liquid, 8mbar at 20 deg.C, 0.6 mPaS at 20 deg.C | CAS: 1330-20-7 | C
8
H
10
Have Questions?
Change view
Quantity:
2.5 L
1 L
5 L
Packaging:
View all
Amber Glass Bottle
Plastic-coated Glass Bottle
Catalog number
FKCX025017
also known as FKCX/0250/17
Price (MYR)
168.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
168.00
BOT
Add to cart
Specifications
Melting Point
-34°C
CAS
1330-20-7
Color
Colorless
Boiling Point
136°C
Quantity
2.5 L
View more
C8H10, CAS Number-1330-20-7, 2.5L, 136 deg.C, Colorless, 106.17g/mol, -34 deg.C, 77264, Amber glass bottle, Liquid, 8mbar at 20 deg.C, 0.6 mPaS at 20 deg.C