Tween™ 80, Pure, Fisher Chemical
Tween™ 80, Pure, Fisher Chemical
Actual product may vary
Tween™ 80, Pure, Fisher Chemical
Tween™ 80, Pure, Fisher Chemical
Tween™ 80, Pure, Fisher Chemical
Have Questions?
Catalog NumberQuantity
FKCT420360
also known as FKCT/4203/60
1 kg
Catalog number FKCT420360
also known as FKCT/4203/60
Price (MYR)
268.00
BOT
Add to cart
Quantity:
1 kg
Price (MYR)
268.00
BOT
Add to cart
1kg, Amber glass bottle
Specifications
ColorAmber
CAS9005-65-6
Physical FormLiquid
pH6
Quantity1 kg
PackagingAmber glass bottle
Molecular Formula(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
MDL NumberMFCD00082107
InChI KeyHDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILESCCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
Molecular Weight (g/mol)604.82
Unit SizeBOT