Check Order Status
Trifluoroacetic Acid, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Trifluoroacetic Acid, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FKCT3256PB05
also known as FKCT/3256/PB05
100 mL
Plastic Coated Glass bottle
Catalog number
FKCT3256PB05
also known as FKCT/3256/PB05
Price (MYR)
269.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (MYR)
269.00
EA
Add to cart
Specifications
pH
0.45
CAS
76-05-1
Melting Point
-15°C
Boiling Point
72°C
Color
Colorless
View more