Toluene, Optima™, Fisher Chemical™
Toluene, Optima™, Fisher Chemical™
Toluene, Optima™, Fisher Chemical™
Toluene, Optima™, Fisher Chemical™
Toluene, Optima™, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Catalog number FKCT230617
also known as FKCT/2306/17
Price (MYR)
141.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
141.00
EA
Add to cart
Specifications
Melting Point-95°C
CAS108-88-3
ColorColorless
Boiling Point111°C
Quantity2.5 L
View more
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only