Check Order Status
Sodium Hydroxide, Certified AR for Analysis, 46-51% Solution, Certified AR, Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide, Certified AR for Analysis, 46-51% Solution, Certified AR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
HDPE plastic bottle
Catalog number
FKCS493017
also known as FKCS/4930/17
Price (MYR)
383.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
383.00
EA
Add to cart
Specifications
pH
13
Boiling Point
145°C
CAS
1310-73-2
Melting Point
12°C
Molecular Formula
HNaO
View more