Check Order Status
Pyridine, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Pyridine, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Packaging:
View all
Plastic-coated Glass Bottle
Catalog number
FKCP7920PB17
also known as FKCP/7920/PB17
Price (MYR)
555.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
555.00
EA
Add to cart
Specifications
pH
8.5
Melting Point
-42°C
CAS
110-86-1
Color
Colorless
Boiling Point
115°C
View more