Check Order Status
Propan-1-ol, for HPLC, Fisher Chemical™
Element Search
Structure Search
Propan-1-ol, for HPLC, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCP748617
also known as FKCP/7486/17
Price (MYR)
266.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
266.00
BOT
Add to cart
Specifications
pH
7
Melting Point
-127°C
CAS
71-23-8
Color
Colorless
Boiling Point
97°C
View more