Check Order Status
Potassium Iodide, Certified AR for Analysis, Fisher Chemical
Element Search
Structure Search
Potassium Iodide, Certified AR for Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FKCP588060
also known as FKCP/5880/60
1 kg
Plastic powder jar
Catalog number
FKCP588060
also known as FKCP/5880/60
Price (MYR)
737.00
Special offer
Ends:
31-Mar-2026
1,279.00
Save 542.00 (42%)
Bottle
Add to cart
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
737.00
Special offer
Ends:
31-Mar-2026
1,279.00
Save 542.00 (42%)
Bottle
Add to cart
Specifications
Boiling Point
1330°C
Melting Point
680°C
CAS
7681-11-0
Color
White
pH
6 to 8
View more