Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
HKO, CAS Number-1310-58-3, potash lye, caustic potash, hydroxyde de potassium, caswell no. 693, potassium hydrate, potassium hydroxide, potasse caustique, potassa, potassium hydroxide solution, potassium hydroxide k oh, 500g, 1320 deg.C, CHEBI:32035, White, 56.11g/mol, KWYUFKZDYYNOTN-UHFFFAOYSA-M
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number FKCP564060
also known as FKCP/5640/60
Price (MYR)
227.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
227.00
EA
Add to cart
Specifications
pH13.5
ColorWhite
Boiling Point1320°C
CAS1310-58-3
Melting Point360°C
View more
HKO, CAS Number-1310-58-3, potash lye, caustic potash, hydroxyde de potassium, caswell no. 693, potassium hydrate, potassium hydroxide, potasse caustique, potassa, potassium hydroxide solution, potassium hydroxide k oh, 500g, 1320 deg.C, CHEBI:32035, White, 56.11g/mol, KWYUFKZDYYNOTN-UHFFFAOYSA-M