▸
Biochemical Reagents
▸
Staining Reagents
▸
Stains and Dyes
▸
Phenolphthalein Solid, Pure, Indicator Grade, Fisher Chemical™
Element Search
Structure Search
Phenolphthalein Solid, Pure, Indicator Grade, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
100 g
Catalog number
FKCP240053
also known as FKCP/2400/53
Price (MYR)
407.00
Bottle
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
407.00
Bottle
Add to cart
Specifications
Melting Point
258°C
CAS
77-09-8
Molecular Formula
C20H14O4
MDL Number
MFCD00005913
InChI Key
KJFMBFZCATUALV-UHFFFAOYSA-N
View more
Phenolphthalein Solid, Pure, Indicator Grade