Check Order Status
1,10-Phenanthroline Monohydrate, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
1,10-Phenanthroline Monohydrate, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
5 g
1 g
100 g
Packaging:
View all
Amber glass bottle
Catalog number
FKCP220044
also known as FKCP/2200/44
Price (MYR)
151.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
151.00
BOT
Add to cart
Specifications
CAS
5144-89-8
Melting Point
97°C
Color
White
Packaging
Amber glass bottle
Molecular Formula
C12H10N2O
View more