Check Order Status
Petroleum Ether 80-100°C, Certified AR for Analysis, N-Hexane <20%, Fisher Chemical™
Element Search
Structure Search
Petroleum Ether 80-100°C, Certified AR for Analysis, N-Hexane <20%, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Catalog number
FKCP184017
also known as FKCP/1840/17
Price (MYR)
202.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
202.00
BOT
Add to cart
Specifications
CAS
64742-49-0
Color
Colorless
Boiling Point
78°C
Quantity
2.5 L
Packaging
Amber Glass Bottle
View more