Petroleum Ether 60-80°C, Certified AR for Analysis, N-Hexane <50%, Fisher Chemical™
Petroleum Ether 60-80&deg;C, Certified AR for Analysis, N-Hexane &lt;50%, Fisher Chemical&trade;
Petroleum Ether 60-80&deg;C, Certified AR for Analysis, N-Hexane &lt;50%, Fisher Chemical&trade;
Petroleum Ether 60-80&deg;C, Certified AR for Analysis, N-Hexane &lt;50%, Fisher Chemical&trade;
Petroleum Ether 60-80°C, Certified AR for Analysis, N-Hexane <50%, Fisher Chemical™
C6H14, CAS Number-64742-49-0, dimethylpropylmethane, unii-49ib0u6mld, isohexane, 1,1-dimethylbutane, 2-methylpentan, 2-methyl pentane, 2-methyl-pentane, iso-hexane, methyl pentane, pentane, 2-methyl, 2.5L, 60 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal Can
Catalog number FKCP180025
also known as FKCP/1800/25
Price (MYR)
1,550.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Can
Request bulk or custom format
Price (MYR)
1,550.00
EA
Add to cart
Specifications
Chemical Name or MaterialPetroleum Ether
CAS64742-49-0
ColorColorless
Boiling Point60°C
Quantity25 L
View more
C6H14, CAS Number-64742-49-0, dimethylpropylmethane, unii-49ib0u6mld, isohexane, 1,1-dimethylbutane, 2-methylpentan, 2-methyl pentane, 2-methyl-pentane, iso-hexane, methyl pentane, pentane, 2-methyl, 2.5L, 60 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178