Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
FKCP112053 also known as FKCP/1120/53 | 500 g |
FGCP112046 also known as FGCP/1120/46 | 25 g |
FGCP112048 also known as FGCP/1120/48 | 100 g |
FKCP112048 also known as FKCP/1120/48 | 100 g |
FGCP112053 also known as FGCP/1120/53 | 500 g |