Check Order Status
Paraffin Liquid, Technical, d=0.88, for Oil Baths, Fisher Chemical™
Element Search
Structure Search
Paraffin Liquid, Technical, d=0.88, for Oil Baths, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Packaging:
Amber glass bottle
Catalog number
FKCP032017
also known as FKCP/0320/17
Price (MYR)
271.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
271.00
EA
Add to cart
Specifications
CAS
8042-47-5
Melting Point
-9°C
Boiling Point
310°C
Packaging
Amber glass bottle
Molecular Formula
MFCD00131611
View more
Paraffin Liquid, Technical, d=0.88, for Oil Baths