Aggressive Adhesive, Self-laminating Vinyl Wrap Around Labels
Aggressive Adhesive, Self-laminating Vinyl Wrap Around Labels
Actual product may vary
Aggressive Adhesive, Self-laminating Vinyl Wrap Around Labels
B-417 High-adhesion, self-laminating vinyl wraps are for tough conditions, including high-humidity environments, small diameter wires and newer wire jacketing materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (LxW)
FKCM449417
also known as FKCM/4494/17
25.40 mm (W) x 25.40 mm (H)
FGCM449417
also known as FGCM/4494/17
25.40 mm (W) x 25.40 mm (H)
Catalog number FKCM449417
also known as FKCM/4494/17
Price (MYR)
280.00
BOT
Add to cart
Dimensions (LxW):
25.40 mm (W) x 25.40 mm (H)
Price (MYR)
280.00
BOT
Add to cart
  • Dependable, long-term adhesion to small diameter wires
  • Will stay adhered in even the most humid environments
  • Strong adhesion to a variety of wire jacketing materials, including Teflon™, silicone, braided and traditional PVC
  • White printable area and a clear overlaminating area that protects the print
  • Ideal for control panels, electrical panels, wire harnesses, datacom and wire and cable ID
Specifications
Certifications/ComplianceUL Recognised to UL969 Labelling and Marking Standard when printed with the Brady Series R4300 and R7950 ribbons. See UL files MH62024 for specific details. Brady customers should specify at purchase if they require UL Recognised B-417 labels. Brady sells both UL Recognised B-417 labels and non-approved labels depending on customer requirements Compliant with Directive 2011/65/EU and 2015/863
Adhesive TypePermanent Acrylic
ColorWhite, Transparent
FinishMatte
FormatDie-cut
MaterialVinyl
Product TypeLabels
SterilityNon-sterile
For Use With (Application)Data and telecommunications labelling;Electrical labelling;Wire and cable labelling
For Use With (Equipment)BMP41;BMP51;BMP53;M511;M410;M510
Quantity260
Temperature Range (English)-40°F to 158°F
Temperature Range (Metric)-40°C to 70°C
Thickness (English)0.004 in
Thickness (Metric)0.107 mm
Wire Gauge16 AWG - 8 AWG
Dimensions (LxW)25.40 mm (W) x 25.40 mm (H)
Length (English)1 in.
Width (English)1 in
Width (Metric)25.40 mm
Width (Metric) Printable Area25.40 mm
Height (English)1 in
Height (Metric)25.40 mm
Alternate Part NumberM4-49-417
Unit SizeBOT