Check Order Status
Methanol, for HPLC-MS, Fisher Chemical™
Element Search
Structure Search
Methanol, for HPLC-MS, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Catalog number
FKCM406217
also known as FKCM/4062/17
Price (MYR)
221.00
BOT
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
221.00
BOT
Add to cart
Specifications
Melting Point
-98°C
CAS
67-56-1
Color
Colorless
Boiling Point
64.7°C
Quantity
2.5 L
View more
Methanol, for HPLC-MS