Check Order Status
Lead, Pure, Metal, Granulated, Fisher Chemical
Element Search
Structure Search
Lead, Pure, Metal, Granulated, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FKCL070053
also known as FKCL/0700/53
500 g
Plastic powder jar
Catalog number
FKCL070053
also known as FKCL/0700/53
Price (MYR)
432.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
432.00
EA
Add to cart
Specifications
Color
Gray
CAS
7439-92-1
Melting Point
327.4°C
Boiling Point
1740°C
Molecular Formula
Pb
View more