Check Order Status
Karl Fischer Aqualine™ Electrolyte AG-H, for Karl Fischer Titration By Coulometry, Fisher Chemical
Element Search
Structure Search
Karl Fischer Aqualine™ Electrolyte AG-H, for Karl Fischer Titration By Coulometry, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FKCK253008
also known as FKCK/2530/08
500 mL
Catalog number
FKCK253008
also known as FKCK/2530/08
Price (MYR)
382.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
382.00
EA
Add to cart
Specifications
Quantity
500 mL
Packaging
Amber glass bottle
Physical Form
Liquid
MDL Number
81734