PureLink™ HiPure Plasmid Filter Midiprep Kit
PureLink™ HiPure Plasmid Filter Midiprep Kit
Actual product may vary
PureLink™ HiPure Plasmid Filter Midiprep Kit
PureLink HiPure Plasmid Filter Midiprep Kit is designed to isolate plasmid DNA of highest purity from 25 to 100 mL of overnight bacterial culture.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FKCK210015
also known as FKCK/2100/15
50 Preps
FGCK210015
also known as FGCK/2100/15
50 Preps
Catalog number FKCK210015
also known as FKCK/2100/15
Price (MYR)
329.00
EA
Add to cart
Quantity:
50 Preps
Price (MYR)
329.00
EA
Add to cart
PureLink HiPure Plasmid Filter Midiprep Kit is designed to isolate plasmid DNA of highest purity from 25 to 100 mL of overnight bacterial culture. The PureLink HiPure Plasmid Filter Kits include HiPure Filter Columns, which combine filtration for clarification of bacterial lysate with anion-exchange resin purification of plasmid DNA, in single unit. The resulting plasmid DNA is of quality equivalent to that obtained by purification with two passes through cesium chloride gradients—the most rigorous method for DNA purification.
  • Purify high-quality DNA in Less than two hours for transfections and other applications with no need for additional steps to remove contaminants like RNA, proteins, or endotoxins
  • Additionally, elimination of phenol, chloroform, ethidium bromide, and cesium chloride from protocol minimizes exposure to and disposal of hazardous materials
PureLink HiPure Plasmid Filter Kits provide:
  • Simple protocols—bacterial lysate clarification without centrifugation
  • Fast results save time by purifying plasmid DNA in less than two hours
  • High yields up to 200 μg for midipreps and up to 850 μg for maxipreps
  • Reliable performance, high-quality plasmid DNA purified with these kits is suitable for wide variety of applications
How it works
  • Addition of a HiPure Filter Column to the PureLink HiPure Plasmid Kits provides rapid clearing of the bacterial lysate without the need for centrifugation
  • Lysate filtration cartridge is integrated into DNA-binding column for a combined step of lysate clarification and plasmid DNA binding directly to the anion-exchange resin
  • Single column wash removes impurities such as RNA, proteins, metabolites, and other low molecular weight molecules; the ultrapure plasmid DNA is then eluted with high-salt buffer
  • DNA is then desalted and concentrated with an isopropanol precipitation step, then collected by centrifugation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeFilter Column
FormatColumn
Isolation TechnologyAnion Exchange Resin
Sample TypeBacterial Culture
Elution Volume5 mL
Final Product TypePlasmid DNA
For Use With (Application)Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions50 Preps
Plasmid<40kb, Low Copy Plasmid, High Copy Plasmid, BAC
Prep Scale100-200 μg (Medium-Scale) Plasmid DNA
Product LinePureLink™
Product TypePlasmid Filter MidiPrep Kit
Quantity50 Preps
PurityTransfection Grade
Shipping ConditionRoom Temperature
TargetPlasmid DNA
System TypePureLink™
Test Time90 min.
Yield350 μg
Unit SizeEA
Contents & Storage
The PureLink™ HiPure Plasmid Filter Kits include Equilibration Buffer (EQ1), Resuspension Buffer (R3), Lysis Buffer (L7), Precipitation Buffer (N3), RNase A, Wash Buffer (W8), Elution Buffer (E4), TE Buffer (TE), and HiPure Filter Columns. Store all components at room temperature upon receipt.