Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
H2O4S, CAS Number-7664-93-9, acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil, 6AMP, 100 deg.C, CHEBI:26836, Colorless, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid, 64589, 98.072
Have Questions?
Catalog NumberQuantity
FKCJ8460C90
also known as FKCJ/8460C/90
6 Ampules
Catalog number FKCJ8460C90
also known as FKCJ/8460C/90
Price (MYR)
643.00
EA
Add to cart
Quantity:
6 Ampules
Request bulk or custom format
Price (MYR)
643.00
EA
Add to cart
Specifications
pH1.5
ColorColorless
CAS7664-93-9
Boiling Point100°C
PackagingAmpoule
View more
H2O4S, CAS Number-7664-93-9, acide sulfurique, acidum sulfuricum, battery acid, dihydrogen sulfate, dipping acid, electrolyte acid, mattling acid, oil of vitriol, sulphuric acid, vitriol brown oil, 6AMP, 100 deg.C, CHEBI:26836, Colorless, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid, 64589, 98.072