Check Order Status
Barium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Element Search
Structure Search
Barium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
Catalog number
FKCJ800408
also known as FKCJ/8004/08
Price (MYR)
464.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
464.00
EA
Add to cart
Specifications
Packaging
HDPE plastic bottle
Physical Form
Liquid
MDL Number
134031
Color
Colorless
pH
1
View more