Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
Hydrochloric Acid, 37%, Certified AR for Analysis, d=1.18, Fisher Chemical™
ClH, CAS Number-7647-01-0, muriatic acid, spirits of salt, hydrogen chloride hcl, hydrogen chloride, hydrochloric acid, chlorwasserstoff, chloorwaterstof, chlorohydric acid, acide chlorhydrique, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Catalog number FKCH1200PB17
also known as FKCH/1200/PB17
Price (MYR)
178.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
178.00
BOT
Add to cart
Specifications
pH1
ColorColorless
CAS7647-01-0
Melting Point-35°C
Boiling Point57°C
View more
ClH, CAS Number-7647-01-0, spirits of salt, hydrogen chloride, hydrochloric acid, hydrogen chloride hcl, chlorohydric acid, chlorwasserstoff, chloorwaterstof, muriatic acid, acide chlorhydrique, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol