Check Order Status
Hydrazinium Sulfate, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Hydrazinium Sulfate, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Change view
Quantity:
250 g
100 g
Catalog number
FKCH070050
also known as FKCH/0700/50
Price (MYR)
161.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
161.00
EA
Add to cart
Specifications
CAS
10034-93-2
Melting Point
254°C
Color
White
pH
1.5
Packaging
Plastic powder jar
View more