n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™
n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™
n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™
n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™
n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Catalog number FKCH010617
also known as FKCH/0106/17
Price (MYR)
456.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
456.00
BOT
Add to cart
Specifications
Melting Point-91°C
CAS142-82-5
ColorColorless
Boiling Point98°C
Quantity2.5 L
View more
  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 197-300nm range, to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Filtered through a sub-micron filter to remove particulates
  • Intended for research use only