Formic Acid, 98-100%, Certified AR for Analysis, Fisher Chemical™
Formic Acid, 98-100%, Certified AR for Analysis, Fisher Chemical™
Formic Acid, 98-100%, Certified AR for Analysis, Fisher Chemical™
Formic Acid, 98-100%, Certified AR for Analysis, Fisher Chemical™
Formic Acid, 98-100%, Certified AR for Analysis, Fisher Chemical™
CH2O2, CAS Number-64-18-6, formylic acid, myrmicyl, acide formique, methanoic acid, hydrogen carboxylic acid, aminic acid, formira, bilorin, collo-bueglatt, formisoton, 2.5L, 101 deg.C, CHEBI:30751, Colorless, 46.02g/mol, BDAGIHXWWSANSR-UHFFFAOYSA-N, formic acid, 8 deg.C, 3297, 46.025 | CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
25 L
Packaging:
View all
Plastic Coated Glass bottle
Catalog number FKCF1900PB17
also known as FKCF/1900/PB17
Price (MYR)
370.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (MYR)
370.00
BOT
Add to cart
Specifications
pH2.1
CAS64-18-6
Melting Point8°C
Boiling Point101°C
ColorColorless
View more
CH2O2, CAS Number-64-18-6, formylic acid, myrmicyl, acide formique, methanoic acid, hydrogen carboxylic acid, aminic acid, formira, bilorin, collo-bueglatt, formisoton, 2.5L, 101 deg.C, CHEBI:30751, Colorless, 46.02g/mol, BDAGIHXWWSANSR-UHFFFAOYSA-N, formic acid, 8 deg.C, 3297, 46.025