Check Order Status
Formic Acid, Pure, about 85%, Fisher Chemical™
Element Search
Structure Search
Formic Acid, Pure, about 85%, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Catalog number
FKCF1750PB17
also known as FKCF/1750/PB17
Price (MYR)
397.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
397.00
EA
Add to cart
Specifications
pH
2.1
CAS
64-18-6
Melting Point
8°C
Boiling Point
101°C
Color
Colorless
View more
Formic Acid, Pure, about 85%