Check Order Status
Ethyl Acetate, for HPLC, Fisher Chemical™
Element Search
Structure Search
Ethyl Acetate, for HPLC, Fisher Chemical™
C4H8O2, CAS Number-141-78-6, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate, -83.5 deg.C, 9171, 88.106, Amber glass bottle, Liquid, 8857, CCOC(-O)C, 3.04, 103mbar at 20 deg.C, 0.45cp at 20 deg.C
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
Amber glass bottle
Catalog number
FKCE090617
also known as FKCE/0906/17
Price (MYR)
289.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
289.00
BOT
Add to cart
Specifications
Packaging
Amber glass bottle
CAS
141-78-6
Vapor Pressure
103mbar at 20°C
Viscosity
0.45cp at 20°C
Vapor Density
3.04
View more
C4H8O2, CAS Number-141-78-6, 2.5L, 75 deg.C, CHEBI:27750, Colorless, 88.11g/mol, XEKOWRVHYACXOJ-UHFFFAOYSA-N, ethyl acetate, -83.5 deg.C, 9171, 88.106, Amber glass bottle, Liquid, 8857, CCOC(-O)C, 3.04, 103mbar at 20 deg.C, 0.45cp at 20 deg.C